Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177041.3 | NP_796015.2 | 3 | 558 | ||
GenBank mRNA | AK044501.1 | - | 567 | |||
AK161929.1 | - | 3 | 602 | |||
BC025817.1 | - | 3 | 511 | 5099127 | ||
BC046769.1 | - | 3 | 558 | 6809196 |