Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177075.4 | NP_796049.1 | 2 | 518 | ||
GenBank mRNA | AK081240.1 | - | 2 | 519 | ||
BC104394.2 | - | 1 | 492 | 40050426 | ||
BC104395.1 | - | 1 | 492 | 40050429 | ||
BY728197.1 | - | 2 | 519 |