Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177082.4 | NP_796056.2 | 3 | 1164 | ||
GenBank mRNA | AJ844914.1 | - | 1 | 851 | ||
AK030745.1 | - | 2 | 983 | |||
AK053093.1 | - | 3 | 1164 |