Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 5, 6 | 886 | ||
GenBank mRNA | AK036709.1 | - | 5, 6 | 887 | ||
AK046672.1 | - | 887 | ||||
AK133242.1 | - | 5, 6 | 1747 | |||
AK155881.1 | - | 5, 6 | 902 | |||
AK170621.1 | - | 5, 6 | 908 | |||
AK171230.1 | - | 5, 6 | 2565 | |||
AK171587.1 | - | 5, 6 | 887 | |||
AK171603.1 | - | 5, 6 | 885 | |||
AK171605.1 | - | 5, 6 | 902 | |||
AY522648.1 | - | 5, 6 | 887 | |||
DQ087179.1 | - | 5, 6 | 639 | |||
DQ087180.1 | - | 6, 7 | 730 |