Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177367.3 | NP_796341.2 | 2 | 641 | ||
GenBank mRNA | AK133821.1 | - | 2 | 733 | ||
AK141078.1 | - | 2 | 620 | |||
AK154061.1 | - | 2 | 622 | |||
AK165864.1 | - | 2 | 585 | |||
BC031425.1 | - | 2 | 581 | 3982927 | ||
BC062157.1 | - | 2 | 589 | 6508223 |