Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177695.4 | NP_808363.3 | 2 | 300 | ||
GenBank mRNA | AK033432.1 | - | 2 | 301 | ||
AY236491.1 | - | 2 | 304 | |||
AY263157.1 | - | 2 | 301 |