Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177695.4 | NP_808363.3 | 5 | 606 | ||
GenBank mRNA | AY236491.1 | - | 5 | 610 | ||
AY263157.1 | - | 5 | 607 |