Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177699.4 | NP_808367.2 | 10 | 1225 | ||
GenBank mRNA | AB041045.1 | - | 10 | 1119 | ||
AK030737.1 | - | 10 | 1187 | |||
BC017144.1 | - | 9 | 1236 | |||
BC060654.1 | - | 10 | 1208 | 30354865 |