Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305043.1 | NP_001291972.1 | 1, 2 | 447 | ||
NM_001305066.1 | NP_001291995.1 | 1, 2 | 447 | |||
NM_004872.4 | NP_004863.2 | 1, 2 | 447 | |||
GenBank mRNA | AF047439.1 | - | 1, 2 | 211 | ||
AF290615.1 | - | 1, 2 | 447 | |||
AK075187.1 | - | 1, 2 | 387 | |||
AK075505.1 | - | 1, 2 | 210 | |||
AK225624.1 | - | 1, 2 | 212 | |||
AX775907.1 | - | 1, 2 | 447 | |||
AX775909.1 | - | 1, 2 | 214 | |||
AY359029.1 | - | 1, 2 | 266 | |||
BC003106.1 | - | 1, 2 | 196 | 3501639 | ||
BC016374.1 | - | 1, 2 | 214 | 4618034 | ||
BC095464.1 | - | 1, 2 | 239 | 30554552 |