Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291190.1 | NP_001278119.1 | 7 | 804 | ||
NM_177710.4 | NP_808378.2 | 8 | 668 | |||
GenBank mRNA | AB099288.1 | - | 8 | 668 | ||
AK155232.1 | - | 7 | 795 | |||
AK170142.1 | - | 7 | 801 | |||
BC141392.1 | - | 8 | 662 | 9056005 | ||
BC141393.1 | - | 8 | 662 | 9056006 |