Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177721.4 | NP_808389.2 | 1 | 3858 | ||
GenBank mRNA | AK083089.1 | - | 1 | 1839 | ||
AK153725.1 | - | 1 | 418 | |||
AK161331.1 | - | 1 | 1561 | |||
BC056759.1 | - | 1 | 3837 | 6518485 |