Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004879.4 | NP_004870.3 | 2, 3 | 308 | ||
XM_011543069.1 | XP_011541371.1 | 3, 4 | 324 | |||
XM_011543070.1 | XP_011541372.1 | 2, 3 | 479 | |||
XM_017018582.1 | XP_016874071.1 | 3, 4 | 324 | |||
GenBank mRNA | AB209820.1 | - | 2, 3 | 265 | ||
AF010313.2 | - | 2, 3 | 180 | |||
AK223029.1 | - | 2, 3 | 237 | |||
AK291951.1 | - | 2, 3 | 217 | |||
AK315841.1 | - | 2, 3 | 224 | |||
BC002390.2 | - | 2, 3 | 174 | 2819783 | ||
BG566671.1 | - | 2, 3 | 282 | |||
BM450796.1 | - | 293 | ||||
DA680745.1 | - | 2, 3 | 308 |