Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colgalt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177756.4 | NP_808424.3 | 2, 3 | 645 | ||
GenBank mRNA | AK052838.1 | - | 2, 3 | 646 | ||
AK076222.1 | - | 2, 3 | 387 | |||
AK085716.1 | - | 2, 3 | 684 | |||
AK134084.1 | - | 2, 3 | 611 | |||
BC068118.1 | - | 2, 3 | 591 | 5706069 |