Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colgalt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177756.4 | NP_808424.3 | 12 | 2355 | ||
GenBank mRNA | AK052838.1 | - | 12 | 2356 | ||
AK076222.1 | - | 12 | 2102 | |||
AK085716.1 | - | 14 | 2547 | |||
AK134084.1 | - | 12 | 2336 | |||
BC068118.1 | - | 12 | 2301 | 5706069 |