Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177771.5 | NP_808439.2 | 7 | 1211 | ||
GenBank mRNA | AK044774.1 | - | 7 | 1322 | ||
AK173029.1 | - | 7 | 1211 | |||
AK192233.1 | - | 2 | 261 | |||
BC025563.1 | - | 7 | 1222 | 5322893 | ||
BC065167.1 | - | 6 | 907 | |||
CB247282.1 | - | 4 | 559 | |||
CF739377.1 | - | 1 | 79 |