Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004882.3 | NP_004873.3 | 3, 4 | 330 | ||
GenBank mRNA | AF098297.1 | - | 3, 4 | 237 | ||
AK292389.1 | - | 3, 4 | 313 | |||
BC015040.1 | - | 3, 4 | 284 | |||
BC021175.1 | - | 3, 4 | 298 | |||
BC031018.1 | - | 3, 4 | 261 | |||
BC038987.1 | - | 3, 4 | 305 | |||
BC046098.2 | - | 3, 4 | 281 | 5495320 | ||
BU596189.1 | - | 3, 4 | 333 | |||
U03644.1 | - | 3, 4 | 269 |