Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178258.5 | NP_839989.2 | 3 | 278 | ||
GenBank mRNA | AF493604.1 | - | 3 | 266 | ||
AJ555484.1 | - | 3 | 272 | |||
AK040641.1 | - | 3 | 287 | |||
AK138913.1 | - | 3 | 279 | |||
AK139024.1 | - | 3 | 309 | |||
BC124998.1 | - | 3 | 268 | 40044741 | ||
BC126872.1 | - | 3 | 278 | 40044756 |