Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163742.1 | NP_001157214.1 | 6 | 1197 | ||
NM_001163743.1 | NP_001157215.1 | 6 | 1298 | |||
NM_178420.3 | NP_848507.2 | 6 | 1286 | |||
GenBank mRNA | AK089260.1 | - | 6 | 1290 | ||
AK141296.1 | - | 6 | 1267 | |||
AK145780.1 | - | 6 | 1308 | |||
AK145988.1 | - | 6 | 1287 | |||
AK166689.1 | - | 6 | 1124 | |||
AK170088.1 | - | 6 | 1241 | |||
BC034204.1 | - | 5 | 1080 | |||
BC050054.1 | - | 6 | 1193 | 30012053 |