Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079847.2 | NP_001073316.1 | 18 | 1637 | ||
NM_001079848.2 | NP_001073317.1 | 17 | 1607 | |||
NM_001079857.2 | NP_001073326.1 | 18 | 1646 | |||
NM_001290445.1 | NP_001277374.1 | 18 | 1640 | |||
NM_001290446.1 | NP_001277375.1 | 19 | 1679 | |||
NM_178712.4 | NP_848827.1 | 19 | 1688 | |||
GenBank mRNA | AF538952.1 | - | 19 | 1653 | ||
AF538955.1 | - | 18 | 1605 | |||
AF538956.1 | - | 19 | 1644 | |||
AF538957.1 | - | 17 | 1572 | |||
AK041291.1 | - | 20 | 1877 | |||
AK046871.1 | - | 18 | 1613 | |||
BC115874.1 | - | 17 | 1582 | 40086356 | ||
BC116644.1 | - | 18 | 1633 | 40086357 |
Set Membership: |
G-protein coupled receptor |