Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178719.5 | NP_848834.2 | 6 | 1450 | ||
GenBank mRNA | AK038466.1 | - | 6 | 1423 | ||
AK041184.1 | - | 6 | 1343 | |||
AK044773.1 | - | 7 | 1883 | |||
AK050546.1 | - | 6 | 1411 | |||
AK080375.1 | - | 6 | 1396 | |||
AK163389.1 | - | 7 | 1552 | |||
BC094597.1 | - | 6 | 1380 | |||
BC137633.1 | - | 5 | 1354 | 8860652 |