Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178719.5 | NP_848834.2 | 6 | 2493 | ||
GenBank mRNA | AK038466.1 | - | 6 | 2465 | ||
AK041184.1 | - | 6 | 2386 | |||
AK050546.1 | - | 6 | 2453 | |||
AK080375.1 | - | 6 | 2440 | |||
BC094597.1 | - | 6 | 2425 |