Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Harbi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178724.4 | NP_848839.2 | 2 | 522 | ||
GenBank mRNA | AK041747.1 | - | 1 | 105 | ||
AK045343.1 | - | 2 | 512 | |||
AK080671.1 | - | 2 | 551 | |||
AK084226.1 | - | 2 | 556 | |||
AK147045.1 | - | 2 | 562 | |||
AK214529.1 | - | 1 | 47 | |||
BC049253.1 | - | 1 | 27 | |||
BC094315.1 | - | 2 | 522 | 5716740 |