Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270411.1 | NP_001257340.1 | 2, 3 | 232 | ||
NM_004900.4 | NP_004891.4 | 2, 3 | 232 | |||
GenBank mRNA | AK024854.1 | - | 2, 3 | 231 | ||
AU098564.1 | - | 2, 3 | 232 | |||
AY743217.1 | - | 2, 3 | 177 |