Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159963.1 | NP_001153435.1 | 2 | 322 | ||
NM_178729.4 | NP_848844.1 | 2 | 322 | |||
GenBank mRNA | AK029957.1 | - | 2 | 270 | ||
AK041497.1 | - | 2 | 294 | |||
AK054009.1 | - | 2 | 292 | |||
AK082931.1 | - | 2 | 292 | |||
AK085100.1 | - | 2 | 295 | |||
AK088067.1 | - | 2 | 289 | |||
AK150664.1 | - | 2 | 282 | |||
AK159744.1 | - | 2 | 311 | |||
AK171361.1 | - | 2 | 276 | |||
AK183264.1 | - | 2 | 172 | |||
BC047214.1 | - | 235 | 6466511 | |||
BC113798.1 | - | 2 | 240 | 40062506 | ||
BY245949.1 | - | 2 | 328 |