Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178730.3 | NP_848845.1 | 9 | 1002 | ||
GenBank mRNA | AK028447.1 | - | 9 | 1004 | ||
AK028587.1 | - | 9 | 997 | |||
AK029032.1 | - | 9 | 964 | |||
AK037173.1 | - | 8 | 846 | |||
BC132629.1 | - | 8 | 852 | 40130906 |