Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011748.2 | NP_035878.1 | 6 | 539 | ||
NM_178733.5 | NP_848848.1 | 5 | 475 | |||
GenBank mRNA | AK028663.1 | - | 5 | 449 | ||
AK032214.1 | - | 5 | 460 | |||
AK045631.1 | - | 6 | 509 | |||
AK220404.1 | - | 3 | 258 | |||
BC066057.1 | - | 5 | 415 | 6833833 |