Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011748.2 | NP_035878.1 | 6 | 1835 | ||
NM_178733.5 | NP_848848.1 | 5 | 1771 | |||
GenBank mRNA | AK028663.1 | - | 5 | 1745 | ||
AK030632.1 | - | 4 | 1641 | |||
AK032214.1 | - | 5 | 1756 | |||
AK045631.1 | - | 6 | 1806 | |||
BC066057.1 | - | 5 | 1711 | 6833833 |