Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170691.1 | NP_001164162.1 | 2, 3 | 277 | ||
NM_178736.5 | NP_848851.3 | 2, 3 | 282 | |||
GenBank mRNA | AK029044.1 | - | 2, 3 | 253 | ||
AK036607.1 | - | 2, 3 | 260 | |||
AK036746.1 | - | 2, 3 | 222 | |||
AK041137.1 | - | 2, 3 | 183 | |||
AK082730.1 | - | 1, 2 | 49 | |||
AK084950.1 | - | 2, 3 | 282 | |||
AK165222.1 | - | 2, 3 | 201 |