Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem229b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170401.1 | NP_001163872.1 | 3 | 1729 | ||
NM_178745.4 | NP_848860.1 | 3 | 1938 | |||
GenBank mRNA | AK033403.1 | - | 3 | 1660 | ||
AK036593.1 | - | 3 | 1697 | |||
AK053623.1 | - | 3 | 1661 | |||
AK149922.1 | - | 1735 | ||||
AK151405.1 | - | 3 | 1732 | |||
AK169759.1 | - | 3 | 1702 | |||
BC057677.1 | - | 3 | 1729 | |||
BC066067.1 | - | 3 | 1938 | 6851142 | ||
BC079613.1 | - | 3 | 1676 | 30361094 |