Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178746.4 | NP_848861.1 | 4 | 426 | ||
GenBank mRNA | AK043402.1 | - | 4 | 414 | ||
AK044735.1 | - | 4 | 416 | |||
BC052361.1 | - | 4 | 426 | 6705789 |