Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178752.3 | NP_848867.2 | 2 | 907 | ||
GenBank mRNA | AK084531.1 | - | 2 | 893 | ||
AK085577.1 | - | 914 | ||||
AK086166.1 | - | 2 | 910 | |||
AK143704.1 | - | 2 | 933 | |||
AK166345.1 | - | 2 | 938 | |||
BC037479.1 | - | 2 | 884 | 4458984 |