Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178759.4 | NP_848874.3 | 9 | 1779 | ||
GenBank mRNA | AK046560.1 | - | 9 | 1778 | ||
AK080042.1 | - | 9 | 1780 | |||
AY376716.1 | - | 9 | 1777 | |||
AY376717.1 | - | 9 | 1777 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC032879.1 | - | 2287 | Chr. - - [-] on Build |