Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114541.1 | NP_001108013.1 | 8 | 1384 | ||
NM_178762.4 | NP_848877.2 | 7 | 1971 | |||
GenBank mRNA | AK031469.1 | - | 4 | 3102 | ||
AK032354.1 | - | 7 | 1963 | |||
AK032762.1 | - | 7 | 1946 | |||
AK044590.1 | - | 4 | 2668 | |||
AK045248.1 | - | 6 | 2041 | |||
AK081584.1 | - | 5 | 1432 | |||
AK085613.1 | - | 6 | 2031 | |||
BC064802.1 | - | 8 | 1430 | 30009464 |