Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256330.4 | XP_005256387.1 | 1, 2 | 109 | ||
XM_011523477.2 | XP_011521779.1 | 1 | 1524 | |||
XM_011523478.2 | XP_011521780.1 | 1 | 1524 | |||
XM_011523480.2 | XP_011521782.1 | 1, 2 | 146 | |||
GenBank mRNA | AB018551.1 | - | 1 | 273 | ||
BC052626.1 | - | 1 | 613 |