Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181517.3 | NP_852658.2 | 1, 2 | 93 | ||
GenBank mRNA | AK039534.1 | - | 1, 2 | 245 | ||
AK053498.1 | - | 1, 2 | 240 | |||
AK079903.1 | - | 1, 2 | 204 | |||
AK148820.1 | - | 1, 2 | 204 | |||
AK169981.1 | - | 1, 2 | 204 | |||
AY316153.1 | - | 1, 2 | 93 |