Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181517.3 | NP_852658.2 | 4 | 470 | ||
GenBank mRNA | AK039534.1 | - | 4 | 622 | ||
AK048477.1 | - | 1 | 1689 | |||
AK148820.1 | - | 4 | 581 | |||
AY316153.1 | - | 4 | 470 | |||
BC053524.1 | - | 1 | 153 |