Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181817.1 | NP_861538.1 | 1 | 1028 | ||
GenBank mRNA | AY288422.1 | - | 1 | 1028 | ||
AY633765.1 | - | 1 | 1028 | |||
BC107269.1 | - | 1 | 1028 | 40052601 | ||
BC107270.1 | - | 1 | 1027 |
Set Membership: |
G-protein coupled receptor |