Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183024.1 | NP_898845.1 | 3 | 404 | ||
GenBank mRNA | AK039072.1 | - | 5 | 292 | ||
AK040404.1 | - | 3 | 460 | |||
BC055329.1 | - | 3 | 404 | 6835616 |