Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183024.1 | NP_898845.1 | 6, 7 | 1241 | ||
GenBank mRNA | AK029114.1 | - | 4, 5 | 737 | ||
AK040404.1 | - | 6, 7 | 1298 | |||
BC055329.1 | - | 6, 7 | 1241 | 6835616 |