Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183027.2 | NP_898848.1 | 3 | 359 | ||
GenBank mRNA | AK028013.1 | - | 3 | 344 | ||
BC054111.1 | - | 3 | 363 | 3154405 | ||
BC098237.1 | - | 3 | 368 | 3825593 | ||
BF715142.1 | - | 3 | 359 |