Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004929.3 | NP_004920.1 | 2, 3 | 339 | ||
GenBank mRNA | AK054881.1 | - | 2, 3 | 337 | ||
AK312491.1 | - | 2, 3 | 337 | |||
BC006478.1 | - | 2, 3 | 327 | 2822391 | ||
BC020864.1 | - | 2, 3 | 337 | 4592497 | ||
HY128922.1 | - | 2, 3 | 340 | |||
S46006.1 | - | 2, 3 | 246 | |||
X06661.1 | - | 2, 3 | 178 |