Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198031.1 | NP_932148.1 | 5 | 416 | ||
GenBank mRNA | AK051775.1 | - | 5 | 428 | ||
AK150149.1 | - | 5 | 440 | |||
AK165503.1 | - | 5 | 439 | |||
AK171491.1 | - | 5 | 425 | |||
AK171804.1 | - | 5 | 431 | |||
BC046615.1 | - | 5 | 416 | |||
BC058566.1 | - | 5 | 416 | 6466057 |