Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159419.1 | NP_001152891.1 | 19 | 2249 | ||
NM_198034.3 | NP_932151.2 | 19 | 2249 | |||
GenBank mRNA | AK034344.1 | - | 4 | 399 | ||
AK171920.1 | - | 19 | 2402 | |||
AY562213.1 | - | 8 | 793 | |||
AY562214.1 | - | 8 | 793 | |||
BC025888.1 | - | 9 | 899 | 5039343 | ||
BC079584.1 | - | 19 | 2249 | 5704672 |