Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC024139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142968.1 | NP_001136440.1 | 8 | 1072 | ||
GenBank mRNA | AK045503.1 | - | 4 | 752 | ||
AK078032.1 | - | 8 | 1060 | |||
AK081549.1 | - | 8 | 1074 | |||
BC060242.1 | - | 8 | 1112 | |||
BC063071.1 | - | 3 | 507 | 5694464 |