Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198617.2 | NP_941019.2 | 1 | 1511 | ||
GenBank mRNA | AK134522.1 | - | 1 | 1508 | ||
AK148882.1 | - | 1 | 1456 | |||
AK167722.1 | - | 1 | 1492 | |||
BC058340.1 | - | 1 | 1453 | 6590443 |