Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198625.1 | NP_941027.1 | 5, 6 | 618 | ||
GenBank mRNA | AK137832.1 | - | 5, 6 | 601 | ||
BC060632.1 | - | 5, 6 | 618 | 6848397 |