Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199018.2 | NP_950183.1 | 5 | 659 | ||
GenBank mRNA | AK133927.1 | - | 5 | 607 | ||
AK149830.1 | - | 5 | 590 | |||
AK150259.1 | - | 5 | 590 | |||
BC062944.1 | - | 5 | 574 | 30357177 | ||
BC078461.1 | - | 4 | 424 |