Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139783.1 | - | 3 | 373 | ||
AK164255.1 | - | 4 | 560 | |||
BC048705.1 | - | 3 | 333 | 6505667 |