Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008398.2 | NP_032424.2 | 5 | 957 | ||
GenBank mRNA | BM950653.1 | - | 1 | 115 | ||
BQ559507.1 | - | 2 | 240 | |||
L23423.1 | - | 5 | 955 |