Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008408.4 | NP_032434.3 | 12 | 1414 | ||
GenBank mRNA | AK030363.1 | - | 12 | 1478 | ||
AK077877.1 | - | 12 | 1438 | |||
AK081547.1 | - | 12 | 1471 | |||
AK087470.1 | - | 12 | 1459 | |||
AK129027.1 | - | 12 | 1452 | |||
AK150640.1 | - | 12 | 1459 | |||
AK151541.1 | - | 12 | 1470 | |||
AK151791.1 | - | 12 | 1470 | |||
AK152242.1 | - | 12 | 1459 | |||
AK153840.1 | - | 12 | 1415 | |||
AK153886.1 | - | 12 | 1515 | |||
AK160037.1 | - | 12 | 1453 | |||
AK160607.1 | - | 12 | 1415 | |||
AK168505.1 | - | 12 | 1460 | |||
AK168937.1 | - | 12 | 1459 | |||
AK169669.1 | - | 12 | 1438 | |||
AX799090.1 | - | 12 | 1457 | |||
BC028897.1 | - | 11 | 1363 | |||
BC037612.1 | - | 12 | 1453 | 5353797 | ||
BC085313.1 | - | 12 | 1443 | 30443880 | ||
L34260.1 | - | 12 | 1457 |